Learn More
Cole-Parmer™ Replacement Impactor for Large Grinding Vial Set (61043-11), Low-chrome Steel
Catalog No. 6871P
Change view
Click to view available options
Material:
Low-chrome Steel
Catalog No. Material
68-71P Low-chrome Steel
1 options
Catalog No. 68-71P Supplier Cole-Parmer™ Supplier No. 6871P
Add to Cart
Add to Cart
Replacement low-chrome steel impactor for (6871) Large low-chrome Grinding Vial Set.
  • Specifically designed for grinding electronic components for RoHS/WEEE compliance testing.
  • Made of low-chrome ASTM 06 steel to eliminate chromium contamination during grinding.
  • Withstands liquid nitrogen temperatures.
  • Autoclavable and reusable.
Specifications
Autoclavable Autoclavable
For Use With (Equipment) 6871 Large Grinding Vial Set (61043-11)
For Use With (Application) For Grinding Electronic Components for RoHS/WEEE Testing, Cr-Free ASTM 06 Steel Eliminates Chromium Contamination
Material Low-chrome Steel
Size Large
WARNING: Reproductive Harm - www.P65Warnings.ca.gov