Learn More
Cole-Parmer Spex VapLock™ Luer Fitting, PP, Straight Bulkhead, Female Luer x 1/4-28 UNF(M) Bottom Sealing Thread; 10/PK
Catalog No. NC2158935 Shop All Cole-Parmer Products
Encompass_Preferred
Choose from fittings constructed from durable polypropylene (PP) or chemically inert PVDF.
This item is not returnable. View return policy
Catalog No. NC2158935 Supplier Cole-Parmer Supplier No. 1202864
Choose from fittings constructed from durable polypropylene (PP) or chemically inert PVDF.