Learn More
Bachem Spexin Trifluoroacetate
SDP
Supplier:  Bachem 4057143.1000
Encompass_Preferred
Spexin Trifluoroacetate, H-Asn-Trp-Thr-Pro-Gln-Ala-Met-Leu-Tyr-Leu-Lys-Gly-Ala-Gln-NH2, 1370290-58-6, C74H114N20O19S, Mr 1619.91, 1mg. Spexin/NPQ human, mouse, bovine. Spexin (SPX, also called NPQ) is a gastro-intestinal peptide hormone. In situ hybridization localized its mRNA to the stomach and esophagus. Synthetic spexin induced dose-dependent contraction of stomach muscle, suggesting that it is involved in digestion. The results obtained by Rucinski et al. indicate that spexin may inhibit adrenocortical cell proliferation.
Catalog No. 50-383-275