Learn More
Cayman Chemical SpGunosIn5O 1thIotrIp 2.7mg
SDP
Catalog No. 502918722
Encompass_Preferred
An isomer of GTP-α-S; binds to tubulin and induces microtubule assembly in cell-free assays at 20 mM
Catalog No. 50-291-8722 Supplier Cayman Chemical Supplier No. 386862.7 MG
An isomer of GTP-α-S; binds to tubulin and induces microtubule assembly in cell-free assays at 20 mM