Learn More
Med Vet International Bips Barium Impregnate Polyethylene Spheres, 4 Small, 1 Large
Catalog No. 502828611
Encompass_Preferred
BIPS, or Barium Impregnated Polyethylene Spheres, provide a range of diagnostic options that reduce the need to undertake exploratory surgery on cats and dogs. Combined with X-rays, spheres of two sizes are used to detect blockages and motility problems in the stomach and intestines. The spheres are administered in food (or by mouth) to avoid the problem of giving liquid barium by stomach tube and syringe. Unlike liquid barium, the small sized BIPS behave in a similar way to food in the gut. Because they can be counted and gut transit times compared with an average speed for healthy animals, the potential for more accurate diagnosis is greatly improved. Packaged in two cards, one with 4 small spheres and the other with 1 large sphere.
Catalog No. 50-282-8611 Supplier Med Vet International Supplier No. PHBIPS
 Restricted Item
Add to Cart
Add to Cart
BIPS, or Barium Impregnated Polyethylene Spheres, provide a range of diagnostic options that reduce the need to undertake exploratory surgery on cats and dogs. Combined with X-rays, spheres of two sizes are used to detect blockages and motility problems in the stomach and intestines. The spheres are administered in food (or by mouth) to avoid the problem of giving liquid barium by stomach tube and syringe. Unlike liquid barium, the small sized BIPS behave in a similar way to food in the gut. Because they can be counted and gut transit times compared with an average speed for healthy animals, the potential for more accurate diagnosis is greatly improved. Packaged in two cards, one with 4 small spheres and the other with 1 large sphere.