Learn More
Chemglass Life Sciences 28/15 O-Ring Ball Joint, Clamp Size: 28, O-Ring Size: 116, Tubing OD: 18.80-19.20mm
SDP
Catalog No. CG12207
Encompass_Preferred
28/15 O-Ring Ball Joint, Clamp Size: 28, O-Ring Size: 116,Tubing OD: 18.80-19.20mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.
Catalog No. CG12207 Supplier Chemglass Life Sciences Supplier No. CG12207
28/15 O-Ring Ball Joint, Clamp Size: 28, O-Ring Size: 116,Tubing OD: 18.80-19.20mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.