Learn More
Wilmad Labglass Spherical NMR Bulb, 18 ul, 10/pack
Catalog No. NC1413206
Encompass_Preferred
Sample volume can be as little as 18 L. 3 different inserts with various shapes (Spherical Bulb, Capillary Tube and Cylindrical Bulb) Outer tube (Precision Thin Walled) must be ordered separately depending on magnetic field strength. Not recommended for variable temperature experiments.
Catalog No. NC1413206 Supplier Wilmad Labglass Supplier No. 529A
Sample volume can be as little as 18 L. 3 different inserts with various shapes (Spherical Bulb, Capillary Tube and Cylindrical Bulb) Outer tube (Precision Thin Walled) must be ordered separately depending on magnetic field strength. Not recommended for variable temperature experiments.