Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Sphericalplate 5D, 3D Cell Culture Plate
SDP

Catalog No. 02681301 Shop All Heidolph North America Products
Change view
Click to view available options
Well Count:
6
24
96
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Count
02-681-301 6
13-876-723 24
01-001-125 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-681-301 Supplier Heidolph™ Supplier No. 036140002
Add to Cart
Add to Cart

The Sphericalplate 5D (SP5D) is a 3D cell culture plate for the formation of uniform, standardized, and size-controlled spheroids in high quality and yield. It also allows you to effortlessly scale up and further translate into diagnostic or clinical applications.

The Sphericalplate 5D (SP5D) is a 3D cell culture plate for the formation of uniform, standardized, and size-controlled spheroids in high quality and yield. It also allows you to effortlessly scale up and further translate into diagnostic or clinical applications. Simultaneously, it increases the reproducibility of subsequent tests, as you always start your experiments with identical initial conditions due to the low size variance of the cultured cell spheroids.

The patented geometry with special coating allows the integration of every single cell into a 3D-agglomerate in a controlled way. This is how uniformly sized spheroids grow without the formation of necrotic cores.

Design:

  • Patented, three-dimensional geometry with rounded bottoms of the microwells, resulting in scaffold-free, regular spheroid formation while keeping the aggregate in the center.
  • The microwell geometry prevents hypoxic damage through aggregate size control.

Ease of Use:

  • There are only three main steps in the SOP and it just takes 24 hours for your 9,000 spheroids to be produced.
  • Step 1) Prepare functional well with medium. Step 2) Add single cell suspension. Step 3) Incubate.
  • Plates are sterilized and have pre-applied anti-adherence coating, making them ready to use out of the box. No pretreatment or centrifugation steps after seeding your cells.
  • Patented surface coating to prevent cell-surface signaling during development and eliminates the need for plate pre-treatment.
  • Special angle of the microwells to allow cells descending in consistent numbers without an additional centrifugation step.

Performance:

  • Spheroid size is controlled by initial seeding density.
  • No sharp edges at the bottom of the plate to reduce surface-induced, non-physiological differentiation.
  • Sharp edges at the top of the microwells to leave no cell behind and no unwanted cell growth outside of the microcavities.
  • Robust non-fouling surface coating featuring low protein-adhesion to allows perfect cell aggregation and prevents surface adhesion.
  • Aggregate size control through the geometric dimensions of the microwells to avoid hypoxia.
  • Selection of COC (cyclic olefin copolymers) material to enable optimal real-time imaging with less background noise.

Applications:

  • Standardization of geometric cluster size for clinical applications.
  • A wide range of cell lines, islet cells, and stem cells have been used on this plate successfully.

24-Well Plates:

  • Standard 24 circular well plate dimension (12 wells have microwells (3D spheroid production), 12 wells do not have microwells (control)).
  • Each well has 750 microwells, allowing the plate to produce up to 9,000 spheroids.
  • Comfortably carry out monolayer comparative studies in the remaining 12 wells of the same plate.

6-Well Plates:

  • Standard 6 square well plate.
  • Expected yield of 3,364 spheroids per well, thus a total yield of 20,184 spheroids per plate.
  • Suitable for scale-up trails and bigger batches.

Specifications

Capacity (English) 0.14 oz.
Closure Included No
Autoclavable Not Autoclavable
Bottom Shape Flat
Capacity (Metric) 4 mL
Color Clear
Disposable Single-use
Material COC
Shape Round
Sterility Sterile
Surface Treatment Non-fouling Coating
Type Cell Culture Dish
Volume (Metric) Working 2 to 4 mL
Well Count 6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.