Learn More
Waters Corp Spherisorb OBD Prep Column, C18, 10 μm, 19 x 250 mm
Catalog No. 502392874 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS1 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS1 packing features low ligand density with a specified carbon load of 6.2%.
  • C18 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • Waters endfitting type
  • OBD prep column format
  • HPLC preparative system
  • LC/MS, LC technique
  • L1 USP classification
  • 6% carbon load
  • Preparative columns product type
  • 1 unit per package
  • 10 μm particle size
  • 80 Å pore size
  • 19 mm inner diameter
  • 250 mm length
  • 2 pH minimum pH range
  • 8 pH maximum pH range
  • 45 °C temperature limit
  • 6000 psi (415 Bar) maximum pressure
  • 220 surface area
Catalog No. 50-239-2874 Supplier Waters Corp Supplier No. 186008857
Spherisorb ODS1 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS1 packing features low ligand density with a specified carbon load of 6.2%.
  • C18 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • Waters endfitting type
  • OBD prep column format
  • HPLC preparative system
  • LC/MS, LC technique
  • L1 USP classification
  • 6% carbon load
  • Preparative columns product type
  • 1 unit per package
  • 10 μm particle size
  • 80 Å pore size
  • 19 mm inner diameter
  • 250 mm length
  • 2 pH minimum pH range
  • 8 pH maximum pH range
  • 45 °C temperature limit
  • 6000 psi (415 Bar) maximum pressure
  • 220 surface area