Learn More
Waters Corp Spherisorb C6 Column, 80 Å, 5 μm, 4.6 x 150 mm
Supplier:  Waters Corp PSS831013RF
Encompass_Preferred
Spherisorb C6 columns are general purpose, silica-based reversed-phase columns that feature a linear-hexyl ligand. These USP L15 columns are designed for applications requiring a less retentive sorbent compared to ODS columns. Each column includes an electronic eConnect column tag for automatic identification, tracking, and digital tracing of use history, optimizing performance when used with the Alliance iS System and Empower software.
  • General purpose silica-based reversed-phase columns with linear-hexyl ligand
  • USP L15 classification for less retentive sorbent needs
  • Electronic tag provides automatic identification and tracking
  • Digitally traces column use history
  • Compatible with all HPLC systems
  • Enhances performance with Alliance iS System and Empower software
Catalog No. 50-239-2555