Learn More
Waters Corp Waters Spherisorb C8 OBD Column, 10 μm, 80Å, 19 mm × 250 mm
Catalog No. 502390824 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Spherisorb C8 OBD Column is designed for preparative chromatography, featuring a 10 μm particle size and 80Å pore size, with dimensions of 19 mm × 250 mm. It is manufactured with patented Optimum Bed Density (OBD) Technology for optimal performance.
  • C8 material for robust separation
  • 10 μm particle size
  • 80Å pore size
  • 19 mm × 250 mm dimensions
  • Manufactured with patented Optimum Bed Density (OBD) technology
Catalog No. 50-239-0824 Supplier Waters Corp Supplier No. 186008859
This Spherisorb C8 OBD Column is designed for preparative chromatography, featuring a 10 μm particle size and 80Å pore size, with dimensions of 19 mm × 250 mm. It is manufactured with patented Optimum Bed Density (OBD) Technology for optimal performance.
  • C8 material for robust separation
  • 10 μm particle size
  • 80Å pore size
  • 19 mm × 250 mm dimensions
  • Manufactured with patented Optimum Bed Density (OBD) technology