Learn More
Waters Corp Spherisorb Cyano (CN) OBD Prep Column, 10 μm, 10 x 250 mm, 80Å Pore Size
Catalog No. 502390475 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb columns are widely referenced HPLC columns in scientific literature, with over 20,000 analytical abstracts published using them. They offer a broad range of validated methods and applications to aid in method development. Spherisorb OBD preparative columns are produced with patented Optimum Bed Density (OBD) Technology, which provides unique separation selectivity to meet chromatographic needs.
  • CN normal phase
  • 10 x 250 mm dimensions
  • 10 μm particle size
  • 3.3 μmol/m² ligand density
  • 3.1% carbon load
  • Not end-capped
  • USP class no. L10
  • 220 m²/g surface area
Catalog No. 50-239-0475 Supplier Waters Corp Supplier No. 186008298
Spherisorb columns are widely referenced HPLC columns in scientific literature, with over 20,000 analytical abstracts published using them. They offer a broad range of validated methods and applications to aid in method development. Spherisorb OBD preparative columns are produced with patented Optimum Bed Density (OBD) Technology, which provides unique separation selectivity to meet chromatographic needs.
  • CN normal phase
  • 10 x 250 mm dimensions
  • 10 μm particle size
  • 3.3 μmol/m² ligand density
  • 3.1% carbon load
  • Not end-capped
  • USP class no. L10
  • 220 m²/g surface area