Learn More
Waters Corp HPLC Column, Spherisorb ODS1, 80Å, 10μm, 4.6 mm × 200 mm
Supplier:  Waters Corp PSS830714RF
Encompass_Preferred
This Waters Spherisorb ODS1 HPLC Column features 80Å pore size, 10μm particle size, and 4.6 mm × 200 mm dimensions. It uses eConnect smart-column technology for enhanced data management and simplified setup. The eConnect tag offers unique product ID, auto-imports data to Empower CDS, and ensures equivalent performance to untagged columns.
  • Second-generation smart-column technology.
  • Electronic device on column body provides unique product ID.
  • NFC technology for convenient product information access.
  • Automated data import and searchable via Empower CDS.
  • Ready for use out of box, simplifying setup & reducing manual errors.
  • Permanently affixed tag reduces compliance, aids auto-detectability by Alliance iS systems.
  • Strategic tag placement enhances detection, minimizes interference.
  • Equivalent performance to untagged columns.
Catalog No. 50-239-2831