Learn More
Waters Corp Spherisorb ODS1 OBD Prep Column, 5 μm, 19 x 250 mm
Catalog No. 502391993 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Spherisorb ODS1 OBD Prep Column is designed with patented Optimum Bed Density (OBD) Technology. It is an ODS1 column with a particle size of 5 μm and dimensions of 19 × 250 mm.
  • Produced with patented Optimum Bed Density (OBD) Technology.
  • Features ODS1 phase.
  • Particle size of 5 μm.
  • Dimensions of 19 × 250 mm.
Catalog No. 50-239-1993 Supplier Waters Corp Supplier No. 186008846
This Spherisorb ODS1 OBD Prep Column is designed with patented Optimum Bed Density (OBD) Technology. It is an ODS1 column with a particle size of 5 μm and dimensions of 19 × 250 mm.
  • Produced with patented Optimum Bed Density (OBD) Technology.
  • Features ODS1 phase.
  • Particle size of 5 μm.
  • Dimensions of 19 × 250 mm.