Learn More
Waters Corp Spherisorb ODS2 Column, 80 Å, 3 μm, 4.6 mm x 150 mm
Catalog No. 502391488 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS2 packing features intermediate ligand density with a specified carbon load of 11.5%. These columns are supplied with industry-standard, Parker-style end fittings. They are manufactured under cGMP guidelines in ISO 9002 certified plants and undergo individual testing, with a test chromatogram provided. Some models are equipped with an electronic eConnect column tag for automatic identification and tracing of use history.
  • Durable, high-efficiency chromatographic columns.
  • Offers exceptional flexibility in optimizing methods and reducing solvent consumption.
  • Facilitates superior performance, reproducibility, and durability in separations.
  • Features a classic pore size of 80Å and a particle size of 3 μm.
Catalog No. 50-239-1488 Supplier Waters Corp Supplier No. PSS832113RF
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS2 packing features intermediate ligand density with a specified carbon load of 11.5%. These columns are supplied with industry-standard, Parker-style end fittings. They are manufactured under cGMP guidelines in ISO 9002 certified plants and undergo individual testing, with a test chromatogram provided. Some models are equipped with an electronic eConnect column tag for automatic identification and tracing of use history.
  • Durable, high-efficiency chromatographic columns.
  • Offers exceptional flexibility in optimizing methods and reducing solvent consumption.
  • Facilitates superior performance, reproducibility, and durability in separations.
  • Features a classic pore size of 80Å and a particle size of 3 μm.