Learn More
Waters Corp Spherisorb OBD Prep Column, ODS2, 80Å, 5 μm, 10 mm x 250 mm
Catalog No. 502391807 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns with intermediate ligand density and a specified carbon load of 11.5%. This column offers exceptional flexibility and less solvent use, providing a reliable and effective chromatographic solution for various applications.
  • Offers a wide range of separation selectivities due to a high-quality bonded phase.
  • Ensures consistent optimal operation, column stability, efficiency, and peak symmetry.
  • Meets stringent requirements for liquid chromatography.
  • Manufactured under cGMP and ISO 9002 requirements for high quality and trustworthy study results.
  • Produced with patented Optimum Bed Density (OBD) Technology.
  • Widely referenced in scientific literature with over 20,000 analytical abstracts.
  • Provides validated methods and applications to aid in method development.
Catalog No. 50-239-1807 Supplier Waters Corp Supplier No. 186008292
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns with intermediate ligand density and a specified carbon load of 11.5%. This column offers exceptional flexibility and less solvent use, providing a reliable and effective chromatographic solution for various applications.
  • Offers a wide range of separation selectivities due to a high-quality bonded phase.
  • Ensures consistent optimal operation, column stability, efficiency, and peak symmetry.
  • Meets stringent requirements for liquid chromatography.
  • Manufactured under cGMP and ISO 9002 requirements for high quality and trustworthy study results.
  • Produced with patented Optimum Bed Density (OBD) Technology.
  • Widely referenced in scientific literature with over 20,000 analytical abstracts.
  • Provides validated methods and applications to aid in method development.