Learn More
Waters Corp Spherisorb OBD Preparative Column, Phenyl, 10 μm, 19 x 250 mm
Catalog No. 502392222 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Spherisorb OBD Preparative Column features a Phenyl phase and 10 μm particle size, designed for high-performance liquid chromatography. It measures 19 × 250 mm and is built with patented Optimum Bed Density (OBD) Technology, ensuring reliable and efficient separations.
  • Widely referenced in scientific literature with over 20,000 analytical abstracts.
  • Offers a broad range of validated methods and applications for method development.
  • Produced using patented optimum bed density technology (US Patent No. 7,399,410 and UK Patent No. GB2408469).
  • Provides unique separation selectivity, suitable for diverse chromatographic requirements.
  • Features a ligand density of 1.7 μmol/m2.
  • Has a carbon load of 2.5%.
  • Not end-capped.
  • USP class No. L11.
  • Surface area of 220 m2/g.
  • Pore size of 80Å.
Catalog No. 50-239-2222 Supplier Waters Corp Supplier No. 186008865
This Spherisorb OBD Preparative Column features a Phenyl phase and 10 μm particle size, designed for high-performance liquid chromatography. It measures 19 × 250 mm and is built with patented Optimum Bed Density (OBD) Technology, ensuring reliable and efficient separations.
  • Widely referenced in scientific literature with over 20,000 analytical abstracts.
  • Offers a broad range of validated methods and applications for method development.
  • Produced using patented optimum bed density technology (US Patent No. 7,399,410 and UK Patent No. GB2408469).
  • Provides unique separation selectivity, suitable for diverse chromatographic requirements.
  • Features a ligand density of 1.7 μmol/m2.
  • Has a carbon load of 2.5%.
  • Not end-capped.
  • USP class No. L11.
  • Surface area of 220 m2/g.
  • Pore size of 80Å.