Learn More
Waters Corp Waters Spherisorb Guard Cartridge, ODS2, 5 μm, 80 Å
Catalog No. 50818678 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge protects analytical columns by removing impurities and suspended solids, extending their lifetime. It features ODS2 packing with intermediate ligand density, manufactured in a cGMP, ISO 9001 certified plant, with each pre-column tested and certified for optimal performance.
  • Silica-based, reversed-phase C18 column with ODS2 packing.
  • Intermediate ligand density and 12% carbon load.
  • Protects analytical columns from impurities, extending lifetime.
  • Designed for biological fluid cleanup/temperature control.
  • pH range of 2 to 8; Max pressure 6000 psi (415 bar).
  • High silanol activity; spherical particle shape.
  • 5 μm particle size; 80 Å pore size.
  • Dimensions: 4.6 mm inner diameter, 10 mm length.
  • HPLC system compatible; USP classification L1.
Catalog No. 50-818-678 Supplier Waters Corp Supplier No. PSS830053
This guard cartridge protects analytical columns by removing impurities and suspended solids, extending their lifetime. It features ODS2 packing with intermediate ligand density, manufactured in a cGMP, ISO 9001 certified plant, with each pre-column tested and certified for optimal performance.
  • Silica-based, reversed-phase C18 column with ODS2 packing.
  • Intermediate ligand density and 12% carbon load.
  • Protects analytical columns from impurities, extending lifetime.
  • Designed for biological fluid cleanup/temperature control.
  • pH range of 2 to 8; Max pressure 6000 psi (415 bar).
  • High silanol activity; spherical particle shape.
  • 5 μm particle size; 80 Å pore size.
  • Dimensions: 4.6 mm inner diameter, 10 mm length.
  • HPLC system compatible; USP classification L1.