Learn More
Waters Corp Spherisorb SCX Column, 80 Å, 5 μm, 4.6 mm × 250 mm
Supplier:  Waters Corp PSS837515RF
Encompass_Preferred
Spherisorb SCX columns contain an anionic silica-based sorbent. This strong cation-exchange sorbent is used to separate positively charged ionic species.
  • Equipped with an electronic eConnect column tag, providing unique column-specific information for automatic identification, tracking, and digital tracing of use history.
  • Offers Cation-Exchange chemistry for ion-exchange separation, built on a spherical silica particle substrate with SCX bonding technology and high silanol activity.
  • Designed for small molecule applications in HPLC systems, operating within a pH range of 2 to 8 and tolerating a maximum pressure of 6000 psi (415 Bar).
  • Key specifications include a particle size of 5 μm, pore size of 80 Å, inner diameter of 4.6 mm, and a length of 250 mm, with Parker-style endfitting.
Catalog No. 50-239-2540