Learn More
Waters Corp Spherisorb SCX OBD Prep Column, 10 μm, 80 Å, 10 mm × 250 mm
Supplier:  Waters Corp 186008303
Encompass_Preferred
Spherisorb SCX columns feature an anionic silica-based sorbent, specifically designed for strong cation-exchange applications. They are used to separate positively charged ionic species.
  • Anionic silica-based sorbent
  • Strong cation-exchange sorbent
  • Designed for separating positively charged ionic species
Catalog No. 50-239-1340