Learn More
Waters Corp Preparative Column, SCX, 10 × 250 mm, 5 μm, 80Å
Catalog No. 502391351 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Spherisorb SCX OBD Preparative Column is engineered using advanced Optimum Bed Density (OBD) Technology to deliver reliable chromatographic performance. These columns are designed for preparative applications, ensuring efficient separations with consistent results.
  • Manufactured with patented Optimum Bed Density (OBD) technology.
  • Features SCX chemistry for robust separations.
  • Dimensions of 10 × 250 mm for preparative scale.
  • Utilizes a 5 μm particle size for optimal resolution.
  • Pore size of 80Å.
  • Surface area of 220 m2/g.
Catalog No. 50-239-1351 Supplier Waters Corp Supplier No. 186008302
The Waters Spherisorb SCX OBD Preparative Column is engineered using advanced Optimum Bed Density (OBD) Technology to deliver reliable chromatographic performance. These columns are designed for preparative applications, ensuring efficient separations with consistent results.
  • Manufactured with patented Optimum Bed Density (OBD) technology.
  • Features SCX chemistry for robust separations.
  • Dimensions of 10 × 250 mm for preparative scale.
  • Utilizes a 5 μm particle size for optimal resolution.
  • Pore size of 80Å.
  • Surface area of 220 m2/g.