Learn More
Waters Corp Spherisorb OBD Preparative Column, Silica, 80Å, 5 μm, 19 × 250 mm
Catalog No. 502392005 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb OBD Preparative Columns are produced with patented Optimum Bed Density (OBD) Technology. The columns feature silica material, 80Å pore size, 5 μm particle size, and 19 × 250 mm dimensions.
  • Optimum bed density technology.
  • Silica material.
  • 80Å pore size.
  • 5 μm particle size.
  • 19 × 250 mm dimensions.
  • Unbonded surface.
Catalog No. 50-239-2005 Supplier Waters Corp Supplier No. 186008851
Spherisorb OBD Preparative Columns are produced with patented Optimum Bed Density (OBD) Technology. The columns feature silica material, 80Å pore size, 5 μm particle size, and 19 × 250 mm dimensions.
  • Optimum bed density technology.
  • Silica material.
  • 80Å pore size.
  • 5 μm particle size.
  • 19 × 250 mm dimensions.
  • Unbonded surface.