Learn More
Spherotech SPHERO ULT.RAIN.PART. 3-3.4UM
Catalog No. NC9029736
Encompass_Preferred
UltraRainbow Fluorescent Particles, 1E7/mL, 3.0-3.4 um, 2 mL
Catalog No. NC9029736 Supplier Spherotech Supplier No. URFP302
UltraRainbow Fluorescent Particles, 1E7/mL, 3.0-3.4 um, 2 mL