Learn More
Alkali Scientific ASI™ Spheroid Dish, 35mm, PS/PE, Sterile, 15/Cs, Low Binding Treatment
SDP
Catalog No. 503028375
Encompass_Preferred
The ASI™ Spheroid Dish is specially designed for efficient spheroid culture and counting. Featuring integrated meshes inside each well, it enables easy identification and accurate quantification of spheroids. The dish ensures seamless collection of spheroids post-experiment, minimizing handling challenges and maximizing efficiency in 3D cell culture research. Mesh Thickness: 137 μm Mesh Pore Size: 200 μm Material: Polystyrene (PS) / Polyethylene (PE) Sterile & Ready to Use Ideal for researchers working with 3D cell cultures, organoids, and tumor spheroids, the ASI™ Spheroid Dish enhances precision and convenience in advanced cell-based studies.
Catalog No. 50-302-8375 Supplier Alkali Scientific Supplier No. SDL35
The ASI™ Spheroid Dish is specially designed for efficient spheroid culture and counting. Featuring integrated meshes inside each well, it enables easy identification and accurate quantification of spheroids. The dish ensures seamless collection of spheroids post-experiment, minimizing handling challenges and maximizing efficiency in 3D cell culture research. Mesh Thickness: 137 μm Mesh Pore Size: 200 μm Material: Polystyrene (PS) / Polyethylene (PE) Sterile & Ready to Use Ideal for researchers working with 3D cell cultures, organoids, and tumor spheroids, the ASI™ Spheroid Dish enhances precision and convenience in advanced cell-based studies.