Learn More
Alkali Scientific ASI™ Spheroid Forming Unit, Sterile, 90/Cs
SDP
Catalog No. 503027613
Encompass_Preferred
Introducing the new Spheroid Forming Unit (SFU) product line.The SFU is specifically designed to support the formation and size scaling of spheroids, enhancing the spheroid generation process.Equipped with a ventilated cap featuring a hydrophobic filter membrane, the SFU ensures efficient gas supply during cell culture. Membrane thickness: 68 μm Pore size: 65 μm Adjustable dial cap for ventilation control 0.2 μm PTFE filter membrane
Catalog No. 50-302-7613 Supplier Alkali Scientific Supplier No. SFU1604
Introducing the new Spheroid Forming Unit (SFU) product line.The SFU is specifically designed to support the formation and size scaling of spheroids, enhancing the spheroid generation process.Equipped with a ventilated cap featuring a hydrophobic filter membrane, the SFU ensures efficient gas supply during cell culture. Membrane thickness: 68 μm Pore size: 65 μm Adjustable dial cap for ventilation control 0.2 μm PTFE filter membrane