Learn More
New England Biolabs, Inc. SphI – 500 units
SDP
Catalog No. 50812101
Encompass
A restriction endonuclease that recognizes the sequence G_CATG^C.
Catalog No. 50-812-101 Supplier New England Biolabs, Inc. Supplier No. R0182S
A restriction endonuclease that recognizes the sequence G_CATG^C.