Learn More
New England Biolabs, Inc. SphI – 2500 units
SDP
Catalog No. 50812100
Encompass
A restriction endonuclease that recognizes the sequence G_CATG^C.
Catalog No. 50-812-100 Supplier New England Biolabs, Inc. Supplier No. R0182M
A restriction endonuclease that recognizes the sequence G_CATG^C.