Learn More
Enzo Life Sciences Dihydrosphingosine, D-erythro (50 mg). CAS: 764-22-7
SDP
Catalog No. 502012114
Encompass_Preferred
Biosynthetic precursor of sphingosine. Inhibitor of protein kinase C (PKC). Blocks phospholipases A2 (PLA2) and the D-sphingosine precursor. Alternative name: D-erythro-Sphingosine, Dihydro-, Sphinganine. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol (25mg/ml, warm) or DMSO (25mg/ml, warm). Long Term Storage: -20°C.
Catalog No. 50-201-2114 Supplier Enzo Life Sciences Supplier No. BMLSL1250050
Biosynthetic precursor of sphingosine. Inhibitor of protein kinase C (PKC). Blocks phospholipases A2 (PLA2) and the D-sphingosine precursor. Alternative name: D-erythro-Sphingosine, Dihydro-, Sphinganine. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol (25mg/ml, warm) or DMSO (25mg/ml, warm). Long Term Storage: -20°C.