Learn More
Cayman Chemical Sphinganine (d18:0), Purity: ≥95%, CAS: 764-22-7, 5 mg
SDP
Catalog No. 502888868
Encompass_Preferred
A precursor of ceramide and sphingosine; can emulate S1P in cell signaling or have opposite intracellular effects
Catalog No. 50-288-8868 Supplier Cayman Chemical Supplier No. 100079455 MG
A precursor of ceramide and sphingosine; can emulate S1P in cell signaling or have opposite intracellular effects