Learn More
Enzo Life Sciences Sphingolactone-24 (1mg). CAS: 881177-99-7
SDP
Supplier:  Enzo Life Sciences ALX270453M001
Encompass_Preferred
Selective and irreversible inhibitor of neutral sphingomyelinase (N-SMase). Purity: ≥95%. Appearance: White waxy solid. Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8576