Learn More
ATCC Campylobacter; jejuni; subsp. jejuni
Catalog No. 502379927
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Campylobacter jejuni subsp. jejuni strain RM3193 was isolated in 1994 from human diarrheal stool. This strain has applications in enteric disease research.
Catalog No. 50-237-9927 Supplier ATCC Supplier No. BAA1234
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Campylobacter jejuni subsp. jejuni strain RM3193 was isolated in 1994 from human diarrheal stool. This strain has applications in enteric disease research.