Learn More
ATCC Sphingomonas paucimobilis
Supplier:  ATCC 10829
Encompass
Sphingomonas paucimobilis strain NRRL B-54 is a bacterium that produces xylose isomerase glucose isomerase.
Catalog No. 50-238-1942