Learn More
Avanti Polar Lipids Sphingomyelin (Milk, Bovine), 475662-40-9, MFCD00132361, 25 mg
Catalog No. NC1526761
Encompass
Synonym: N-(tricosanoyl)-sphing-4-enine-1-phosphocholine, Formula Weight: 785.034, Purity >99%. As a major constituent of cell membranes, sphingomyelin is found at particularly high concentrations in the membranes of nerve cells (in the myelin sheaths) and red blood cells.
Catalog No. NC1526761 Supplier Avanti Polar Lipids Supplier No. 860063C25MG
Synonym: N-(tricosanoyl)-sphing-4-enine-1-phosphocholine, Formula Weight: 785.034, Purity >99%. As a major constituent of cell membranes, sphingomyelin is found at particularly high concentrations in the membranes of nerve cells (in the myelin sheaths) and red blood cells.