Learn More
Medchemexpress LLC Sphingomyelin | 85187-10-6 | ≥99.0% | 493.6 g/mol | 25 MG
SDP
Catalog No. 5000438599
Encompass_Preferred
Sphingomyelin is a eukaryotic sphingolipid that is a major constituent of cell membranes and is particularly abundant in the myelin sheath that surrounds neuronal axons. It plays an important role in cell processes, the regulation of inflammatory responses, and signal transduction. Sphingomyelin metabolism is associated with various central nervous system diseases and Niemann-Pick disease.
  • Endogenous metabolite
  • Plays an important role in cell processes, the regulation of inflammatory responses, and signal transduction.
  • Metabolism is associated with various central nervous system diseases and Niemann-Pick disease.
Catalog No. 50-004-38599 Supplier Medchemexpress LLC Supplier No. 25MGHY113498
Sphingomyelin is a eukaryotic sphingolipid that is a major constituent of cell membranes and is particularly abundant in the myelin sheath that surrounds neuronal axons. It plays an important role in cell processes, the regulation of inflammatory responses, and signal transduction. Sphingomyelin metabolism is associated with various central nervous system diseases and Niemann-Pick disease.
  • Endogenous metabolite
  • Plays an important role in cell processes, the regulation of inflammatory responses, and signal transduction.
  • Metabolism is associated with various central nervous system diseases and Niemann-Pick disease.