Learn More
Medchemexpress LLC Sphingomyelin | 85187-10-6 | ≥99.0% | 50 MG
SDP
Catalog No. 5000437065
Encompass_Preferred
Sphingomyelin is a eukaryotic sphingolipid and a major constituent of cell membranes, particularly abundant in the myelin sheath that surrounds neuronal axons. It plays an important role in cell processes, the regulation of inflammatory responses, and signal transduction. Sphingomyelin metabolism is associated with various central nervous system diseases and Niemann-Pick disease.
  • Major constituent of cell membranes
  • Abundant in the myelin sheath
  • Plays a role in cell processes
  • Regulates inflammatory responses
  • Involved in signal transduction
  • Metabolism associated with central nervous system diseases and Niemann-Pick disease
Catalog No. 50-004-37065 Supplier Medchemexpress LLC Supplier No. HY11349850MG
Sphingomyelin is a eukaryotic sphingolipid and a major constituent of cell membranes, particularly abundant in the myelin sheath that surrounds neuronal axons. It plays an important role in cell processes, the regulation of inflammatory responses, and signal transduction. Sphingomyelin metabolism is associated with various central nervous system diseases and Niemann-Pick disease.
  • Major constituent of cell membranes
  • Abundant in the myelin sheath
  • Plays a role in cell processes
  • Regulates inflammatory responses
  • Involved in signal transduction
  • Metabolism associated with central nervous system diseases and Niemann-Pick disease