Learn More
Echelon Biosciences Research Labs SPHINGOMYELIN BEADS 1 mL
SDP
Catalog No. 50720690
Encompass
Sphingomyelin Beads are composed of agarose with 10 nanomoles of bound sphingomyelin per 1mL of beads, enough for several protein binding experiments. Each 1 mL order includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-690 Supplier Echelon Biosciences Research Labs Supplier No. PB0SM1ML
Sphingomyelin Beads are composed of agarose with 10 nanomoles of bound sphingomyelin per 1mL of beads, enough for several protein binding experiments. Each 1 mL order includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable