Learn More
Medchemexpress LLC Sphingomyelin (d18:1/12:0) | 474923-21-2 | 646.92 | C35H71N2O6P | 5mg
SDP
Catalog No. 5000270337
Encompass_Preferred
Sphingomyelin (d18 1/12 0) is a polar lipid with the activity of regulating cell signaling Sphingomyelin (d18 1/12 0) plays an important role in cell membranes affecting membrane fluidity and stability Sphingomyelin (d18 1/12 0) is also involved in lipid metabolism and has the potential to promote neuronal cell growth Sphingomyelin (d18 1/12 0) plays a key role in the function and development of the nervous system
Catalog No. 50-002-70337 Supplier Medchemexpress LLC Supplier No. HYW9236425MG
Sphingomyelin (d18 1/12 0) is a polar lipid with the activity of regulating cell signaling Sphingomyelin (d18 1/12 0) plays an important role in cell membranes affecting membrane fluidity and stability Sphingomyelin (d18 1/12 0) is also involved in lipid metabolism and has the potential to promote neuronal cell growth Sphingomyelin (d18 1/12 0) plays a key role in the function and development of the nervous system