Learn More
Medchemexpress LLC Sphingomyelin Syntha 50ug
SDP
Catalog No. 5000180757
Encompass_Preferred
The sphingomyelin synthase 2 (SGMS2) protein plays a crucial role in plasma membrane sphingomyelin synthesis It catalyzes the reversible transfer of the phosphocholine moiety in sphingomyelin biosynthesis to form ceramide phosphocholine Sphingomyelin Synthase 2/SGMS2 Protein Human (HEK293 Fc) is the recombinant human-derived Sphingomyelin Synthase 2/SGMS2 protein expressed by HEK293 with N-mFc labeled tag
Catalog No. 50-001-80757 Supplier Medchemexpress LLC Supplier No. HYP7363250UG
The sphingomyelin synthase 2 (SGMS2) protein plays a crucial role in plasma membrane sphingomyelin synthesis It catalyzes the reversible transfer of the phosphocholine moiety in sphingomyelin biosynthesis to form ceramide phosphocholine Sphingomyelin Synthase 2/SGMS2 Protein Human (HEK293 Fc) is the recombinant human-derived Sphingomyelin Synthase 2/SGMS2 protein expressed by HEK293 with N-mFc labeled tag