Learn More
Enzo Life Sciences Sphingomyelinase (Staphylococcus aureus) (1x96wells). CAS: 9031-54-3
SDP
Catalog No. 502011932
Encompass_Preferred
Alternative name: Sphingomyelin cholinephosphohydrolase. Formulation: Liquid. Supplied in 50% glycerol/0.25 M phosphate buffer. Source: Isolated from Staphylococcus aureus. Long Term Storage: +4°C.
Catalog No. 50-201-1932 Supplier Enzo Life Sciences Supplier No. BMLSE1080050
Alternative name: Sphingomyelin cholinephosphohydrolase. Formulation: Liquid. Supplied in 50% glycerol/0.25 M phosphate buffer. Source: Isolated from Staphylococcus aureus. Long Term Storage: +4°C.