Learn More
TARGETMOL CHEMICALS INC Sphingosine-1-phosphate 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T215001MG
Encompass_Preferred
Sphingosine-1-phosphate (S1P) is an agonist for S1P1-5 receptors and a ligand for GPR3, GPR6, and GPR12. As an intracellular second messenger, it mobilizes Ca2+ and acts as an extracellular ligand for G-protein coupled receptors. This important lipid mediator is generated from sphingosine or other membrane phospholipids. Purity 98.66%
Catalog No. 50-287-9613