Learn More
Medchemexpress LLC Sphingosine-1-phosphate | 26993-30-6 | 99.5% | 379.47 | 10 MG
SDP
Catalog No. 5000346944
Encompass_Preferred
Sphingosine-1-phosphate (S1P) is an agonist of S1P1-5 receptors and a ligand of GPR3, GPR6, and GPR12. It functions as an intracellular second messenger and mobilizes Ca2+ as an extracellular ligand for G protein-coupled receptors. S1P is a crucial lipid mediator generated from Sphingomyelin or other membrane phospholipids and stimulates DNA synthesis, cell proliferation, and migration.
  • Agonist for S1P1-5 receptors
  • Ligand for GPR3, GPR6, and GPR12
  • Functions as an intracellular second messenger
  • Mobilizes Ca2+ as an extracellular ligand
  • Crucial lipid mediator
  • Stimulates DNA synthesis, cell proliferation, and migration
  • High purity
  • Solid appearance, white to yellow color
  • For research use only
Catalog No. 50-003-46944 Supplier Medchemexpress LLC Supplier No. HY10849650MG
Sphingosine-1-phosphate (S1P) is an agonist of S1P1-5 receptors and a ligand of GPR3, GPR6, and GPR12. It functions as an intracellular second messenger and mobilizes Ca2+ as an extracellular ligand for G protein-coupled receptors. S1P is a crucial lipid mediator generated from Sphingomyelin or other membrane phospholipids and stimulates DNA synthesis, cell proliferation, and migration.
  • Agonist for S1P1-5 receptors
  • Ligand for GPR3, GPR6, and GPR12
  • Functions as an intracellular second messenger
  • Mobilizes Ca2+ as an extracellular ligand
  • Crucial lipid mediator
  • Stimulates DNA synthesis, cell proliferation, and migration
  • High purity
  • Solid appearance, white to yellow color
  • For research use only