Learn More
Medchemexpress LLC Sphingosine-1-phosphate | 26993-30-6 | 99.5% | 379.47 | 50 MG
SDP
Catalog No. 5000345221
Encompass_Preferred
Sphingosine-1-phosphate (S1P) acts as an agonist for S1P1-5 receptors and serves as a ligand for GPR3, GPR6, and GPR12. It functions as an intracellular second messenger and triggers Ca2+ mobilization as an extracellular ligand for G protein-coupled receptors. S1P is a crucial lipid mediator derived from Sphingomyelin or other membrane phospholipids.
  • Promotes DNA synthesis.
  • Promotes cell proliferation.
  • Promotes migration.
  • Acts as an intracellular second messenger.
  • Triggers Ca2+ mobilization.
Catalog No. 50-003-45221 Supplier Medchemexpress LLC Supplier No. HY108496100MG
Sphingosine-1-phosphate (S1P) acts as an agonist for S1P1-5 receptors and serves as a ligand for GPR3, GPR6, and GPR12. It functions as an intracellular second messenger and triggers Ca2+ mobilization as an extracellular ligand for G protein-coupled receptors. S1P is a crucial lipid mediator derived from Sphingomyelin or other membrane phospholipids.
  • Promotes DNA synthesis.
  • Promotes cell proliferation.
  • Promotes migration.
  • Acts as an intracellular second messenger.
  • Triggers Ca2+ mobilization.