Learn More
Selleck Chemical LLC Sphingosine-1-phosphate E1999-1MG
SDP
Catalog No. 5000039204
Encompass_Preferred
Sphingosine-1-phosphate (S1P) is an agonist of S1P1-5 receptors and a ligand of GPR3 GPR6 and GPR12 Sphingosine-1-phosphate is an intracellular second messenger and mobilizes Ca2 as an extracellular ligand for G protein-coupled receptors Sphingosine-1-phosphate is an important lipid mediator generated from Sphingomyelin or other membrane phospholipids
Catalog No. 50-000-39204 Supplier Selleck Chemical LLC Supplier No. E19991MG
Sphingosine-1-phosphate (S1P) is an agonist of S1P1-5 receptors and a ligand of GPR3 GPR6 and GPR12 Sphingosine-1-phosphate is an intracellular second messenger and mobilizes Ca2 as an extracellular ligand for G protein-coupled receptors Sphingosine-1-phosphate is an important lipid mediator generated from Sphingomyelin or other membrane phospholipids