Learn More
Medchemexpress LLC Sphingosine-1-phosphate (S1P) | 26993-30-6 | 99.5% | 379.47 | 25 MG
SDP
Catalog No. 5000347303
Encompass_Preferred
Sphingosine-1-phosphate (S1P) is an agonist of S1P1-5 receptors and a ligand of GPR3, GPR6 and GPR12. It functions as an intracellular second messenger and mobilizes Ca2+ as an extracellular ligand for G protein-coupled receptors. This lipid mediator stimulates DNA synthesis, cell proliferation, and migration.
  • Agonist of S1P1-5 receptors and a ligand of GPR3, GPR6, and GPR12.
  • Functions as an intracellular second messenger.
  • Mobilizes Ca2+ as an extracellular ligand for G protein-coupled receptors.
  • Important lipid mediator.
  • Stimulates DNA synthesis, cell proliferation, and migration.
  • For research use only.
Catalog No. 50-003-47303 Supplier Medchemexpress LLC Supplier No. 25MGHY108496
Sphingosine-1-phosphate (S1P) is an agonist of S1P1-5 receptors and a ligand of GPR3, GPR6 and GPR12. It functions as an intracellular second messenger and mobilizes Ca2+ as an extracellular ligand for G protein-coupled receptors. This lipid mediator stimulates DNA synthesis, cell proliferation, and migration.
  • Agonist of S1P1-5 receptors and a ligand of GPR3, GPR6, and GPR12.
  • Functions as an intracellular second messenger.
  • Mobilizes Ca2+ as an extracellular ligand for G protein-coupled receptors.
  • Important lipid mediator.
  • Stimulates DNA synthesis, cell proliferation, and migration.
  • For research use only.