Learn More
Echelon Biosciences Research Labs D-Erythro-Sphingosine 10 mg
SDP
Catalog No. 50720761
Encompass
Sphingosine is formed through ceramidase catalyzed hydrolysis of ceramide. It is a potent inhibitor of protein kinase C (IC50 = 1-3 M). Sphingosine is phosphorylated by sphingosine kinase to the bioactive lipid sphingosine 1-phosphate This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-761 Supplier Echelon Biosciences Research Labs Supplier No. S100010MG
Sphingosine is formed through ceramidase catalyzed hydrolysis of ceramide. It is a potent inhibitor of protein kinase C (IC50 = 1-3 M). Sphingosine is phosphorylated by sphingosine kinase to the bioactive lipid sphingosine 1-phosphate This item is non returnable