Learn More
Selleck Chemical LLC Sphingosine-25mg
SDP
Catalog No. 502389891
Encompass_Preferred
Sphingosine (2-amino-4-trans-octadecene-1,3-diol, C18 Sphingosine) is an 18-carbon amino alcohol with an unsaturated hydrocarbon chain, which forms a primary part of sphingolipids, a class of cell membrane lipids that include sphingomyelin, an important phospholipid.
Catalog No. 50-238-9891 Supplier Selleck Chemical LLC Supplier No. S221025MG
Sphingosine (2-amino-4-trans-octadecene-1,3-diol, C18 Sphingosine) is an 18-carbon amino alcohol with an unsaturated hydrocarbon chain, which forms a primary part of sphingolipids, a class of cell membrane lipids that include sphingomyelin, an important phospholipid.