Learn More
Echelon Biosciences Research Labs SPHINGOSINE ARRAYS 5 pack
SDP
Catalog No. 50720779
Encompass
Sphingo Arrays are 4 x 5 cm hydrophobic membranes that have been spotted with a concentration gradient of eight different sphingolipids. They are used to determine the relative degree of binding of your protein of interest towards one or more of these lipids in a simple protein lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-779 Supplier Echelon Biosciences Research Labs Supplier No. S60015PK
Sphingo Arrays are 4 x 5 cm hydrophobic membranes that have been spotted with a concentration gradient of eight different sphingolipids. They are used to determine the relative degree of binding of your protein of interest towards one or more of these lipids in a simple protein lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable