Learn More
Echelon Biosciences Research Labs SPHINGOSINE BEADS 1 mL
SDP
Catalog No. 50720781
Encompass
Sphingo Beads are composed of agarose with 10 nanomoles of bound Sphingosine per mL of beads, enough for several protein binding experiments. Each 1 mL order includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-781 Supplier Echelon Biosciences Research Labs Supplier No. S61001ML
Sphingo Beads are composed of agarose with 10 nanomoles of bound Sphingosine per mL of beads, enough for several protein binding experiments. Each 1 mL order includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable