Learn More
Enzo Life Sciences Sphingosine, D-erythro, (100mg), CAS Number: 123-78-4
SDP
Catalog No. NC1677502
Encompass_Preferred
Potent and selective inhibitor of protein kinase C. Common component of cell membranes, often as phosphatidyl conjugates. Is a potent (IC.+,K+-ATPase, CTP:phosphocholine cytidylyltransferase, calmodulin-dependent enzymes, binding of factor VII to tissue factor, binding of thyrotropin releasing hormone to its receptor and activation of EGF receptor kinase, phospholipase D and casein kinase II. Alternative Name: D(+)-erythro-1,3-Dihydroxy-2-amino-4- trans-octadecene. Formula: C18H37NO2. MW: 299.5. CAS Number: 123-78-4. MI: 14: 8747. Purity: ≥98% (TLC). Appearance: White to off-white solid. Solubility: Soluble in 100% ethanol (25mg/ml warm) or DMSO (25mg/ml warm). Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C
Catalog No. NC1677502 Supplier Enzo Life Sciences Supplier No. BMLEI1550100
Potent and selective inhibitor of protein kinase C. Common component of cell membranes, often as phosphatidyl conjugates. Is a potent (IC.+,K+-ATPase, CTP:phosphocholine cytidylyltransferase, calmodulin-dependent enzymes, binding of factor VII to tissue factor, binding of thyrotropin releasing hormone to its receptor and activation of EGF receptor kinase, phospholipase D and casein kinase II. Alternative Name: D(+)-erythro-1,3-Dihydroxy-2-amino-4- trans-octadecene. Formula: C18H37NO2. MW: 299.5. CAS Number: 123-78-4. MI: 14: 8747. Purity: ≥98% (TLC). Appearance: White to off-white solid. Solubility: Soluble in 100% ethanol (25mg/ml warm) or DMSO (25mg/ml warm). Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C