Learn More
Medchemexpress LLC Sphingosine (d14:1) | 24558-60-9 | MFCD08703027 | 99.7% | 243.39 | C14H29NO2 | 1 MG
SDP
Catalog No. 5000227441
Encompass_Preferred
Sphingosine (d14:1) is a bioactive sphingolipid that functions as a protein kinase C (PKC) inhibitor supplied as a high-purity solid for research applications in biochemical and cell biology studies.
  • Bioactive sphingolipid that inhibits protein kinase C (PKC).
  • High purity (99.7%) suitable for analytical and research use.
  • Available as a solid in a 1 MG pack for small-scale experiments.
  • Powder stable at -20°C for long-term storage (typical handling recommended).
  • Molecular formula C14H29NO2 and molecular weight 243.39.
Catalog No. 50-002-27441 Supplier Medchemexpress LLC Supplier No. HY1184421MG
Sphingosine (d14:1) is a bioactive sphingolipid that functions as a protein kinase C (PKC) inhibitor supplied as a high-purity solid for research applications in biochemical and cell biology studies.
  • Bioactive sphingolipid that inhibits protein kinase C (PKC).
  • High purity (99.7%) suitable for analytical and research use.
  • Available as a solid in a 1 MG pack for small-scale experiments.
  • Powder stable at -20°C for long-term storage (typical handling recommended).
  • Molecular formula C14H29NO2 and molecular weight 243.39.